Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249274.1 | NP_001236203.1 | 3 - 4 | 481 | 98 | |
GenBank mRNA | AF095445.1 | - | 3 - 4 | 481 | 98 | |
BT096859.1 | - | 3 - 4 | 417 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Glycine max Probe spans exons |