Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P5CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251224.1 | NP_001238153.1 | 7 - 8 | 929 | 89 | |
GenBank mRNA | AY492005.1 | - | 7 - 8 | 929 | 89 | |
FM999729.1 | - | 7 - 8 | 929 | 89 | ||
FM999730.1 | - | 7 - 8 | 929 | 89 | ||
FN564570.1 | - | 7 - 8 | 929 | 89 | ||
FN564571.1 | - | 7 - 8 | 929 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |