Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU019571.1 | - | 1 - 2 | 311 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Glycine max Probe spans exons |