Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004122.hg.1 |
TC03000208.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004122.hg.1 |
ENST00000301825.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004122.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004122.hg.1 |
ENST00000456402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004122.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149997.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149997.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149997.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149997.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149997.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149997.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149997.hg.1 |
TC0300007117.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007117.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000301825.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000439533.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000445129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
ENST00000456402.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
NM_001248.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
NM_001291960.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
NM_001291961.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300150001.hg.1 |
TC0300007117.hg.1 |
Clariom D TC |