Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs00159041_m1
See other MAP2 GE Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs00159041_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_002374.3
NP_002365.3
9 - 10
4771
59
XM_005246555.1
XP_005246612.1
10 - 11
5136
59
XM_005246565.1
XP_005246622.1
6 - 7
4425
59
XM_005246566.1
XP_005246623.1
6 - 7
4413
59
XM_006712532.1
XP_006712595.1
10 - 11
5133
59
XM_011511186.1
XP_011509488.1
10 - 11
5043
59
XM_011511187.1
XP_011509489.1
9 - 10
4937
59
XM_011511190.1
XP_011509492.1
9 - 10
4940
59
XM_011511192.1
XP_011509494.1
9 - 10
4940
59
XM_011511195.1
XP_011509497.1
8 - 9
4691
59
XM_011511196.2
XP_011509498.1
9 - 10
4878
59
XM_011511197.2
XP_011509499.1
9 - 10
4875
59
XM_017004111.1
XP_016859600.1
9 - 10
4937
59
XM_017004112.1
XP_016859601.1
9 - 10
4890
59
XM_017004113.1
XP_016859602.1
9 - 10
4782
59
XM_017004114.1
XP_016859603.1
9 - 10
4890
59
XM_017004115.1
XP_016859604.1
8 - 9
4682
59
XM_017004117.1
XP_016859606.1
8 - 9
4679
59
XM_017004118.1
XP_016859607.1
9 - 10
4797
59
XM_017004119.1
XP_016859608.1
9 - 10
4887
59
XM_017004120.1
XP_016859609.1
9 - 10
4887
59
XM_017004122.1
XP_016859611.1
9 - 10
4794
59
XM_017004123.1
XP_016859612.1
9 - 10
4878
59
XM_017004124.1
XP_016859613.1
9 - 10
4785
59
XM_017004125.1
XP_016859614.1
9 - 10
4875
59
XM_017004127.1
XP_016859616.1
9 - 10
4875
59
XM_017004128.1
XP_016859617.1
8 - 9
4676
59
XM_017004130.1
XP_016859619.1
6 - 7
4862
59
XM_017004131.1
XP_016859620.1
6 - 7
4425
59
XM_017004132.1
XP_016859621.1
6 - 7
4413
59
XM_017004133.1
XP_016859622.1
6 - 7
4425
59
XM_017004134.1
XP_016859623.1
6 - 7
4413
59
XM_017004135.1
XP_016859624.1
6 - 7
4859
59
XM_017004136.1
XP_016859625.1
6 - 7
4422
59
GenBank mRNA
M25668.1
-
3 - 4
3044
59
U01828.1
-
9 - 10
4771
59
U89330.1
-
6 - 7
4511
59
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Homo sapiens
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene MAP2
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
MicroArray Probes:
ABI
Array Name
Probe IDs
Targets
Target Type
ABI Human Genome Survey Microarray Version 2.0
131819
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0