Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180421.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180421.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180421.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180421.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180421.hg.1 |
TC0400011706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
ENST00000274024.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011706.hg.1 |
NM_000134.3 |
Refseq |