Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000411817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000411817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000411817.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000461533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000461533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
ENST00000461533.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003420.hg.1 |
TC03000181.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003420.hg.1 |
ENST00000264741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003420.hg.1 |
ENST00000422441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003420.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003419.hg.1 |
TC03000181.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003419.hg.1 |
ENST00000264741.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003419.hg.1 |
ENST00000422441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003419.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149194.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149194.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149194.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149194.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149194.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149194.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149194.hg.1 |
TC0300007051.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149191.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149191.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149191.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149191.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149191.hg.1 |
TC0300007051.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000411817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000411817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000411817.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000461533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000461533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
ENST00000461533.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007051.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149193.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149193.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149193.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149193.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149193.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149193.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149193.hg.1 |
TC0300007051.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300149192.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149192.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149192.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149192.hg.1 |
ENST00000264741.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149192.hg.1 |
ENST00000422441.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149192.hg.1 |
NM_002207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149192.hg.1 |
TC0300007051.hg.1 |
Clariom D TC |