Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001715.hg.1 |
TC07000111.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001715.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001715.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001715.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146914.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146914.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146914.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146914.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146914.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146914.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146914.hg.1 |
TC0700006783.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000262067.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
ENST00000466195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006783.hg.1 |
NM_014399.3 |
Refseq |