Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010735.hg.1 |
ENST00000613889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011252.hg.1 |
ENST00000420733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000270257.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000391951.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591607.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000591747.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000592351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000592351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
ENST00000592351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_001007270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011764.hg.1 |
NM_024707.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009460.hg.1 |
ENST00000451718.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007718.hg.1 |
ENST00000422502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000342595.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000378220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
ENST00000486842.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007034.hg.1 |
NM_022567.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009460.hg.1 |
ENST00000451718.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007718.hg.1 |
ENST00000422502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010735.hg.1 |
ENST00000613889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011252.hg.1 |
ENST00000420733.1 |
Ensembl |