Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009296.hg.1 |
ENST00000325279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009296.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009296.hg.1 |
TC16000658.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009295.hg.1 |
ENST00000325279.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009295.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009295.hg.1 |
TC16000658.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000258157.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000565743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000567410.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
ENST00000569472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_001303451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008656.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
ENST00000325279.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164705.hg.1 |
TC1600008656.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600164701.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164701.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164701.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164701.hg.1 |
ENST00000564159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164701.hg.1 |
ENST00000564996.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164701.hg.1 |
NM_024731.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164701.hg.1 |
TC1600008656.hg.1 |
Clariom D TC |