Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000221972.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000444740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
ENST00000597454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_001783.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008166.hg.1 |
NM_021601.3 |
Refseq |