Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007346.hg.1 |
ENST00000447106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002159.hg.1 |
TC06000240.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002159.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002160.hg.1 |
TC06000240.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002160.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149887.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149887.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149887.hg.1 |
TC0600007346.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007346.hg.1 |
ENST00000447106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007346.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149888.hg.1 |
ENST00000447106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149888.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149888.hg.1 |
ENST00000447106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149888.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149888.hg.1 |
TC0600007346.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149886.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149886.hg.1 |
NR_045612.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149886.hg.1 |
TC0600007346.hg.1 |
Clariom D TC |