Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000712.hg.1 |
ENST00000252321.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000712.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000712.hg.1 |
TC12000071.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000713.hg.1 |
ENST00000252321.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000713.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000713.hg.1 |
TC12000071.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000711.hg.1 |
ENST00000252321.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000711.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12000711.hg.1 |
TC12000071.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006574.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145319.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145319.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145319.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145319.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145319.hg.1 |
TC1200006574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200145322.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145322.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145322.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145322.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145322.hg.1 |
TC1200006574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200145320.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145320.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145320.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145320.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145320.hg.1 |
TC1200006574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200145321.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145321.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145321.hg.1 |
ENST00000252321.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200145321.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145321.hg.1 |
TC1200006574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200145317.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145317.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145317.hg.1 |
TC1200006574.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200145318.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145318.hg.1 |
NM_002234.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200145318.hg.1 |
TC1200006574.hg.1 |
Clariom D TC |