Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000338353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000354422.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000435676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000441649.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
ENST00000456900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016276.hg.1 |
NM_001017961.4 |
Refseq |