Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000460204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000460204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001918.hg.1 |
TC07000125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001918.hg.1 |
ENST00000222573.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001918.hg.1 |
ENST00000537992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001918.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147296.hg.1 |
TC0700006844.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000222573.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000460204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000460204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000477859.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000478974.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
ENST00000537992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006844.hg.1 |
NM_002214.2 |
Refseq |