Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042406.1 | NP_001035865.1 | 7 - 8 | 957 | 65 | |
NM_001287741.1 | NP_001274670.1 | 6 - 7 | 861 | 65 | ||
NM_001287746.1 | NP_001274675.1 | 8 - 9 | 1053 | 65 | ||
NM_001287753.1 | NP_001274682.1 | 4 - 5 | 648 | 65 | ||
NM_019036.2 | NP_061909.2 | 8 - 9 | 1047 | 65 | ||
GenBank mRNA | AF131827.1 | - | 3 - 4 | 351 | 65 | |
AK055075.1 | - | 8 - 9 | 938 | 65 | ||
AK127587.1 | - | 8 - 9 | 1018 | 65 | ||
AK293856.1 | - | 6 - 7 | 722 | 65 | ||
AK294210.1 | - | 4 - 5 | 600 | 65 | ||
BC024194.2 | - | 7 - 8 | 838 | 4818781 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |