Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017890.4 | NP_060360.3 | 51 - 52 | 9512 | 80 | |
NM_152564.4 | NP_689777.3 | 51 - 52 | 9437 | 80 | ||
XM_005250800.3 | XP_005250857.1 | 51 - 52 | 9818 | 80 | ||
XM_005250801.4 | XP_005250858.1 | 51 - 52 | 15688 | 80 | ||
XM_011516848.2 | XP_011515150.1 | 51 - 52 | 9501 | 80 | ||
XM_011516849.2 | XP_011515151.1 | 51 - 52 | 9426 | 80 | ||
XM_011516850.2 | XP_011515152.1 | 49 - 50 | 9144 | 80 | ||
XM_011516851.2 | XP_011515153.1 | 31 - 32 | 6404 | 80 | ||
XM_011516852.2 | XP_011515154.1 | 31 - 32 | 6442 | 80 | ||
XM_011516854.2 | XP_011515156.1 | 27 - 28 | 5600 | 80 | ||
XM_017013109.1 | XP_016868598.1 | 50 - 51 | 9309 | 80 | ||
XM_017013110.1 | XP_016868599.1 | 44 - 45 | 8302 | 80 | ||
XM_017013111.1 | XP_016868600.1 | 31 - 32 | 6674 | 80 | ||
XM_017013112.1 | XP_016868601.1 | 28 - 29 | 5816 | 80 | ||
GenBank mRNA | AB011104.1 | - | 12 - 13 | 2246 | 80 | |
AJ608772.1 | - | 51 - 52 | 9504 | 80 | ||
AJ608773.1 | - | 51 - 52 | 9429 | 80 | ||
AY223814.1 | - | 51 - 52 | 9512 | 80 | ||
AY223815.1 | - | 51 - 52 | 9437 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |