Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000548918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000548918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
ENST00000616161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011567.hg.1 |
NR_106998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000261226.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000548918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000548918.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
ENST00000616161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NM_020698.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011567.hg.1 |
NR_106998.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189592.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189592.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189592.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189592.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189592.hg.1 |
TC1200011567.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200189591.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189591.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189591.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189591.hg.1 |
ENST00000551457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189591.hg.1 |
ENST00000552239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189591.hg.1 |
NM_001301036.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189591.hg.1 |
TC1200011567.hg.1 |
Clariom D TC |