Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019940.hg.1 |
TC03001154.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019940.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019940.hg.1 |
ENST00000412055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019940.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019941.hg.1 |
TC03001154.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019941.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019941.hg.1 |
ENST00000412055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03019941.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010223.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174554.hg.1 |
TC0300010223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174553.hg.1 |
TC0300010223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
ENST00000295984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
ENST00000411976.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
ENST00000412055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
NM_207351.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174552.hg.1 |
TC0300010223.hg.1 |
Clariom D TC |