Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_001110781_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7911594 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7911596 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7911595 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7911593 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7911592 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7911591 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
217122_PM_s_at |
- |
- |
Affymetrix Human Genome U219 Array |
11724093_s_at |
- |
- |
Affymetrix Human Genome U219 Array |
11724093_PM_s_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032241.hg.1 |
TC01006344.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032241.hg.1 |
ENST00000234800.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032241.hg.1 |
ENST00000378662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032241.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032241.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006526.hg.1 |
ENST00000607013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006526.hg.1 |
ENST00000607013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032242.hg.1 |
TC01006344.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032242.hg.1 |
ENST00000234800.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032242.hg.1 |
ENST00000378662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032242.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032242.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032240.hg.1 |
TC01006344.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032240.hg.1 |
ENST00000234800.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032240.hg.1 |
ENST00000378662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032240.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006526.hg.1 |
ENST00000607013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006526.hg.1 |
ENST00000607013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241066.hg.1 |
TC0100018388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241068.hg.1 |
TC0100018388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100241067.hg.1 |
TC0100018388.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000480991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000481276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000611123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000614300.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
ENST00000617444.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001110781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018388.hg.1 |
NM_001290264.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
215169_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
215169_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
217122_PM_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
217122_s_at |
- |
- |
Human Genome U133A 2.0 Array |
215169_PM_at |
- |
- |
Human Genome U133A 2.0 Array |
217122_s_at |
- |
- |
Human Genome U133A 2.0 Array |
215169_at |
- |
- |
Human Genome U133A 2.0 Array |
217122_PM_s_at |
- |
- |
Human Genome U95 Set |
41243_PM_at |
- |
- |
Human Genome U95 Set |
41243_at |
- |
- |
Human Genome U95 Set |
67594_PM_at |
- |
- |
Human Genome U95 Set |
67594_at |
- |
- |
Human Genome U133 Set |
217122_s_at |
- |
- |
Human Genome U133 Set |
215169_PM_at |
- |
- |
Human Genome U133 Set |
217122_PM_s_at |
- |
- |
Human Genome U133 Set |
215169_at |
- |
- |