Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000622969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000622969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000622969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229523.hg.1 |
TC0100017083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000622969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000622969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000622969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017083.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229525.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229525.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229525.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229525.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229525.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229525.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229525.hg.1 |
TC0100017083.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000617070.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000617991.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000623597.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
ENST00000623893.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
NM_001164522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
NM_001304839.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
NM_177403.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229519.hg.1 |
TC0100017083.hg.1 |
Clariom D TC |