Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NR_003921_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8106021 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8106022 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177557 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8106023 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177555 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104630 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104631 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177556 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104629 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104628 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8106020 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8177558 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8106019 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8177554 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8104627 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
224419_PM_x_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
224421_PM_x_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
224418_PM_x_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
217123_PM_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11760089_x_at |
- |
- |
Affymetrix Human Genome U219 Array |
11760089_PM_x_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000457791.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000457791.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000523548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000523548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
ENST00000523548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001189.hg.1 |
TC05000111.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001189.hg.1 |
ENST00000418902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001189.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500146997.hg.1 |
TC0500006935.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000415808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000450213.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000457791.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000457791.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000523548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000523548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
ENST00000523548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007722.hg.1 |
NR_003922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000418902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000423301.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
ENST00000430902.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006935.hg.1 |
NR_003921.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
224418_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
224418_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
224421_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
224419_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
224419_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
224421_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
217123_PM_x_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
217123_x_at |
- |
- |
Human Genome U133A 2.0 Array |
217123_PM_x_at |
- |
- |
Human Genome U133A 2.0 Array |
217123_x_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw509716 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224421_x_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
217123_x_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224418_x_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
224419_x_at10 |
- |
- |
Human Genome U95 Set |
62823_PM_at |
- |
- |
Human Genome U95 Set |
62823_at |
- |
- |
Human Genome U133 Set |
217123_x_at |
- |
- |
Human Genome U133 Set |
217123_PM_x_at |
- |
- |
Human Genome U133 Set |
224419_PM_x_at |
- |
- |
Human Genome U133 Set |
224418_PM_x_at |
- |
- |
Human Genome U133 Set |
224421_PM_x_at |
- |
- |
Human Genome U133 Set |
224421_x_at |
- |
- |
Human Genome U133 Set |
224418_x_at |
- |
- |
Human Genome U133 Set |
224419_x_at |
- |
- |