Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017277.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017277.hg.1 |
TC10001365.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017275.hg.1 |
ENST00000287139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017275.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017275.hg.1 |
TC10001365.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017276.hg.1 |
ENST00000287139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017276.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017276.hg.1 |
TC10001365.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010921.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174367.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174367.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174367.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174367.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174367.hg.1 |
ENST00000414871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174367.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174367.hg.1 |
TC1000010921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174368.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174368.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174368.hg.1 |
ENST00000287139.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174368.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174368.hg.1 |
TC1000010921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174369.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174369.hg.1 |
NM_018055.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174369.hg.1 |
TC1000010921.hg.1 |
Clariom D TC |