Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008242.hg.1 |
ENST00000216139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008242.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008242.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008242.hg.1 |
TC22000436.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008241.hg.1 |
ENST00000216139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008241.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008241.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008241.hg.1 |
TC22000436.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000527761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000527761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000527761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000527761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000527761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000527761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007826.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
TC2200007826.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155698.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
TC2200007826.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
NM_001097.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
ENST00000216139.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
ENST00000529621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
ENST00000533930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155699.hg.1 |
NM_001097.2 |
Refseq |