Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005645.hg.1 |
ENST00000216014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005645.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005645.hg.1 |
TC22000301.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005644.hg.1 |
ENST00000216014.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005644.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005644.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005644.hg.1 |
TC22000301.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151781.hg.1 |
TC2200007356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151781.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151781.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151781.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151781.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151781.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151781.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151780.hg.1 |
TC2200007356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151780.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151780.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151780.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151780.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151780.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151780.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000409006.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
ENST00000471268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007356.hg.1 |
NM_016657.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151782.hg.1 |
TC2200007356.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151782.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151782.hg.1 |
NM_006855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151782.hg.1 |
ENST00000216014.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151782.hg.1 |
NM_006855.3 |
Refseq |