Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059892.hg.1 |
TC01003949.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059892.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059892.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008808.hg.1 |
ENST00000566803.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008808.hg.1 |
ENST00000566803.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008808.hg.1 |
ENST00000566803.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059893.hg.1 |
TC01003949.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059893.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059893.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059894.hg.1 |
TC01003949.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059894.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059894.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006524.hg.1 |
ENST00000614297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006524.hg.1 |
ENST00000614297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006524.hg.1 |
ENST00000614297.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006668.hg.1 |
ENST00000561950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006668.hg.1 |
ENST00000561950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006668.hg.1 |
ENST00000561950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233407.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233407.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233407.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233407.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233407.hg.1 |
TC0100017666.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000486755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
ENST00000498158.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017666.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233408.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233408.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233408.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233408.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233408.hg.1 |
TC0100017666.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233409.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233409.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233409.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233409.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233409.hg.1 |
TC0100017666.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100233410.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233410.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233410.hg.1 |
ENST00000344517.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100233410.hg.1 |
NM_012089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100233410.hg.1 |
TC0100017666.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500006668.hg.1 |
ENST00000561950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006668.hg.1 |
ENST00000561950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006668.hg.1 |
ENST00000561950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006524.hg.1 |
ENST00000614297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006524.hg.1 |
ENST00000614297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006524.hg.1 |
ENST00000614297.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008808.hg.1 |
ENST00000566803.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008808.hg.1 |
ENST00000566803.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008808.hg.1 |
ENST00000566803.2 |
Ensembl |