Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463611 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463623 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463630 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463645 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463609 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255357 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463648 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463604 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255377 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463602 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255397 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255374 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463617 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463654 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463627 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463646 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463633 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463635 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255363 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463631 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463640 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463652 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463653 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255385 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255395 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255386 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463606 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463608 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463610 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15894422 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463647 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255360 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463625 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255393 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255415 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255373 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463621 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463601 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463632 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463655 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463644 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463607 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463629 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255396 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255362 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25463649 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255379 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05255391 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
164663 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
42777 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
267763 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
794406 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
191838 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
303335 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
12862 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
AF230386.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
AF230387.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
AK222729.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
AK290270.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
BC016866.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
BC017017.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
BT006675.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
NM_007028.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
142562 |
X81006.1 |
genbank_mrna_human |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P38283 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_32_P38286 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_32_P38283 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P416732 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
23593 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
16132 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
38261 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P117729 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P117729 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
8514 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P01641068 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_14_P117729 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31937659 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P37897250 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P15951106 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P38283 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_19_P00812159 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_32_P38283 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463644 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463632 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463655 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463621 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463625 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463649 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463652 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463647 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463628 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463645 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463630 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463635 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255357 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255385 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255399 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463627 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463629 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463633 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255395 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255393 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463640 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463636 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15894422 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255414 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463623 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463651 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463632 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463628 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463636 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463645 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463635 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255395 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255357 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463621 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463649 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463647 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463623 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255393 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463652 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463630 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463629 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463633 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463640 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463644 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463627 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463651 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463625 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255385 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463655 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255399 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05255414 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25463654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15894422 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_32_P38286 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P416732 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_32_P38283 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P117729 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255405 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255385 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255390 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255389 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255411 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255414 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255408 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255399 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255394 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255401 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255387 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255393 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255397 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255409 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P05255407 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15894421 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P05255413 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P05255415 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P05255414 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15894422 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P17818831 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P117729 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P117729 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P416732 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_32_P38286 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_32_P38283 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
100016 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
97810 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P15952425 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01641063 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P17818831 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P117729 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P17818828 |
- |
- |
Agilent Human 1B Microarray G4111A |
A_32_P832191 |
- |
- |
Agilent Human 1B Microarray G4111A |
A_32_P38289 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255380 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255387 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255399 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255351 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255384 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255393 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255378 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255409 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255370 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255357 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255359 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255376 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15894417 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255408 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255397 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255391 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15894419 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255404 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15894415 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255401 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255405 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255414 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255382 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255360 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255353 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15894414 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255407 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255355 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255367 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255374 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255362 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255411 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05255349 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P117729 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P17818831 |
- |
- |