Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002224.3 | NP_002215.2 | 27 - 28 | 3743 | 65 | |
XM_011514577.2 | XP_011512879.1 | 26 - 27 | 3383 | 65 | ||
XM_017010832.1 | XP_016866321.1 | 28 - 29 | 3572 | 65 | ||
GenBank mRNA | AB209739.1 | - | 18 - 19 | 2530 | 65 | |
D26351.1 | - | 27 - 28 | 3754 | 65 | ||
U01062.1 | - | 27 - 28 | 3555 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Homo sapiens Probe spans exons |