Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017489.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017489.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017489.hg.1 |
TC14001563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017488.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017488.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017488.hg.1 |
TC14001563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176825.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176825.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176825.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176825.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176825.hg.1 |
TC1400010394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
ENST00000392590.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010394.hg.1 |
NM_145701.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176826.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176826.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176826.hg.1 |
ENST00000336219.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176826.hg.1 |
NM_017955.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176826.hg.1 |
TC1400010394.hg.1 |
Clariom D TC |