Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002289.hg.1 |
TC6_cox_hap2000134.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002289.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002289.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001809.hg.1 |
TC6_mann_hap4000112.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001809.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001809.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002005.hg.1 |
TC6_mcf_hap5000114.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002005.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002005.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002263.hg.1 |
TC6_qbl_hap6000124.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002263.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002263.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002063.hg.1 |
TC6_ssto_hap7000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002063.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002063.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016840.hg.1 |
TC06001499.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016840.hg.1 |
ENST00000437089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016840.hg.1 |
ENST00000453195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016840.hg.1 |
ENST00000454678.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016840.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016840.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002010.hg.1 |
TC6_mcf_hap5000114.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002010.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002010.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000947.hg.1 |
TC6_apd_hap1000070.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000947.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000947.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002219.hg.1 |
TC6_dbb_hap3000123.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002219.hg.1 |
NM_001242783.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002219.hg.1 |
NM_003449.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176461.hg.1 |
TC0600011357.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000416596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000418026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000434785.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000437089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000453195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000454678.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000480999.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011357.hg.1 |
ENST00000487829.1 |
Ensembl |