Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000552052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000552052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000552052.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000553161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000553161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
ENST00000553161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011532.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011532.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011532.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011532.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011532.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011532.hg.1 |
TC12000874.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166038.hg.1 |
TC1200008900.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000455836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000552052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000552052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000552052.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000553161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000553161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
ENST00000553161.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_001034025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008900.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166036.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166036.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166036.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166036.hg.1 |
ENST00000261735.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166036.hg.1 |
ENST00000546477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200166036.hg.1 |
NM_006817.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200166036.hg.1 |
TC1200008900.hg.1 |
Clariom D TC |