Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003577.2 | NP_003568.2 | 2 | 951 | 102 | |
GenBank mRNA | AB011076.2 | - | 2 | 952 | 102 | |
AY606112.1 | - | 2 | 951 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592552 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592554 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592546 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592547 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27984814 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27984811 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07592553 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3294217 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2324 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P33141 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2323 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 37932 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P105491 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P105491 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 9886 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3294217 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3294217 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592546 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984814 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592552 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592546 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984814 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27984811 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07592552 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P33141 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P105491 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07592552 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07592550 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07592553 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07592549 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07592554 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07592545 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P07592546 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403353 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403357 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07592546 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403356 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403354 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403347 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07592545 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403348 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403349 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07592553 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403340 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403358 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403351 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07592554 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07592551 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403352 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403355 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403341 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07592552 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16403350 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P105491 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P105491 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P33141 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 102599 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 7722 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32807346 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592575 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403357 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403355 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592545 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592546 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403379 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403376 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403371 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592550 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403346 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592595 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592586 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592591 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592549 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403364 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592572 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592593 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592580 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592587 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592553 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592579 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403383 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403352 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592590 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592589 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592552 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592554 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403353 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16403368 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592582 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07592573 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_003577_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7931554 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7931555 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7931553 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208275_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738674_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738673_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738674_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11738673_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR10011769.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011769.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011769.hg.1 | TC10000950.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000009451.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009451.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009451.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009451.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009451.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009451.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164590.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164590.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164590.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000164590.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000164590.hg.1 | TC1000009451.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000009451.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009451.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009451.hg.1 | ENST00000304477.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009451.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009451.hg.1 | NM_003577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009451.hg.1 | NM_003577.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 208275_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 208275_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 208275_x_at | - | - |
Human Genome U133A 2.0 Array | 208275_PM_x_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003577_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw527241 | - | - |
Human Genome U95 Set | 31393_PM_r_at | - | - |
Human Genome U95 Set | 31393_r_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208275_x_at2 | - | - |
Human Genome U133 Set | 208275_x_at | - | - |
Human Genome U133 Set | 208275_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044681 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044681 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 149070 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768793 | AB011076.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_4507858-S | - | - |
Illumina HumanRef-8 v2 Expression BeadChip | GI_14411-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0000610053 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0000610053 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 2900424 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_14411 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1768793 | AY606112.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768793 | NM_003577.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768793 | AB011076.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768793 | AY606112.1 | human_transcriptome |