Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012434.hg.1 |
TC03000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012434.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012434.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012433.hg.1 |
TC03000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012433.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012433.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162519.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162519.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162519.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162519.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162519.hg.1 |
TC0300008787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
ENST00000296271.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008787.hg.1 |
NM_000539.3 |
Refseq |