Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055512.hg.1 |
TC01003608.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055512.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055512.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226181.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226181.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226181.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226181.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226181.hg.1 |
TC0100016631.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
ENST00000367558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016631.hg.1 |
NM_002928.3 |
Refseq |