Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000467226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000467226.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002703.hg.1 |
TC02004956.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002703.hg.1 |
ENST00000334056.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002703.hg.1 |
ENST00000379579.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002703.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002703.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002703.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000449210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000467226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000467226.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
ENST00000491044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016423.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
ENST00000334056.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
ENST00000379579.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
ENST00000439646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
NM_001008779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
NM_001142634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
NM_182756.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200221427.hg.1 |
TC0200016423.hg.1 |
Clariom D TC |