Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005625.hg.1 |
ENST00000257934.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005625.hg.1 |
ENST00000552462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005625.hg.1 |
ENST00000552671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005625.hg.1 |
NM_012291.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12005625.hg.1 |
TC12000434.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000257934.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000549154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000552462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000552671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
NM_012291.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000257934.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000549154.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000552462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
ENST00000552671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
NM_012291.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200155134.hg.1 |
TC1200007707.hg.1 |
Clariom D TC |