Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009622.hg.1 |
TC01000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009622.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009622.hg.1 |
ENST00000495703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009622.hg.1 |
ENST00000539779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01009622.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100157981.hg.1 |
TC0100008121.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000372090.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000460057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000471337.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000477731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
ENST00000495703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008121.hg.1 |
NM_025077.3 |
Refseq |