Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122956.1 | NP_001116428.1 | 7 - 8 | 806 | 87 | |
NM_001284315.1 | NP_001271244.1 | 5 - 6 | 618 | 87 | ||
NM_014063.6 | NP_054782.2 | 7 - 8 | 806 | 87 | ||
GenBank mRNA | AK027367.1 | - | 7 - 8 | 735 | 87 | |
AK293151.1 | - | 6 - 7 | 669 | 87 | ||
AK293279.1 | - | 6 - 7 | 626 | 87 | ||
AK302037.1 | - | 7 - 8 | 790 | 87 | ||
BC011677.2 | - | 7 - 8 | 729 | 4299867 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |