Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278185.1 | NP_001265114.1 | 7 - 8 | 1085 | 161 | |
NM_015601.3 | NP_056416.2 | 7 - 8 | 1085 | 161 | ||
NM_022079.2 | NP_071362.1 | 7 - 8 | 1085 | 161 | ||
XM_011539592.2 | XP_011537894.1 | 8 - 9 | 1159 | 161 | ||
XM_011539593.2 | XP_011537895.1 | 8 - 9 | 1159 | 161 | ||
XM_011539594.2 | XP_011537896.1 | 9 - 10 | 1245 | 161 | ||
XM_011539595.2 | XP_011537897.1 | 9 - 10 | 1295 | 161 | ||
XM_011539596.2 | XP_011537898.1 | 8 - 9 | 1173 | 161 | ||
XM_011539597.2 | XP_011537899.1 | 8 - 9 | 1159 | 161 | ||
XM_017016041.1 | XP_016871530.1 | 7 - 8 | 1087 | 161 | ||
GenBank mRNA | AB046813.1 | - | 4 - 5 | 939 | 161 | |
AK292809.1 | - | 7 - 8 | 965 | 161 | ||
AY221963.1 | - | 7 - 8 | 1022 | 161 | ||
AY650033.1 | - | 7 - 8 | 1022 | 161 | ||
BC039600.1 | - | 7 - 8 | 995 | 5212048 | 161 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |