Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006584.hg.1 |
ENST00000253799.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006584.hg.1 |
ENST00000452774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006584.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006584.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006584.hg.1 |
TC17000539.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157844.hg.1 |
TC1700007918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000253799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
ENST00000452774.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_001158.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007918.hg.1 |
NM_009590.3 |
Refseq |