Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002034.hg.1 |
TC02000163.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002034.hg.1 |
ENST00000380320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002034.hg.1 |
ENST00000544143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002034.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002036.hg.1 |
TC02000163.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002036.hg.1 |
ENST00000380320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002036.hg.1 |
ENST00000544143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002036.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002035.hg.1 |
TC02000163.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002035.hg.1 |
ENST00000380320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002035.hg.1 |
ENST00000544143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002035.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148155.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148155.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148155.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148155.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148155.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148155.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148155.hg.1 |
TC0200007052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200148157.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148157.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148157.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148157.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148157.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148157.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148157.hg.1 |
TC0200007052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200148156.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148156.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148156.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148156.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148156.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148156.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148156.hg.1 |
TC0200007052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200148154.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148154.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148154.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148154.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148154.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148154.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148154.hg.1 |
TC0200007052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000380320.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
ENST00000433140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007052.hg.1 |
NM_007046.3 |
Refseq |