Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017497.hg.1 |
TC09001405.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017497.hg.1 |
ENST00000342112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017497.hg.1 |
ENST00000375063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017497.hg.1 |
ENST00000375064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017497.hg.1 |
ENST00000375066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017497.hg.1 |
NM_018421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017500.hg.1 |
TC09001405.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017500.hg.1 |
ENST00000342112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017500.hg.1 |
ENST00000375063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017500.hg.1 |
ENST00000375064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017500.hg.1 |
ENST00000375066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017500.hg.1 |
NM_018421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017499.hg.1 |
TC09001405.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017499.hg.1 |
ENST00000342112.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017499.hg.1 |
ENST00000375063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017499.hg.1 |
ENST00000375064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017499.hg.1 |
ENST00000375066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09017499.hg.1 |
NM_018421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000342112.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000375063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000375064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000375066.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000465784.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NM_001267571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NM_001267572.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NM_018421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NR_051978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000342112.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000375063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000375064.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000375066.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
ENST00000465784.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NM_001267571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NM_001267572.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NM_018421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
NR_051978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900174761.hg.1 |
TC0900010971.hg.1 |
Clariom D TC |