Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000342.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000342.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000342.hg.1 |
TC14000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000343.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000343.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000343.hg.1 |
TC14000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000338.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000338.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000338.hg.1 |
TC14000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000339.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000339.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000339.hg.1 |
TC14000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000341.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000341.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14000341.hg.1 |
TC14000069.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400144611.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144611.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144611.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144611.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144611.hg.1 |
TC1400006533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400144607.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144607.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144607.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144607.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144607.hg.1 |
TC1400006533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400144608.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144608.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144608.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144608.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144608.hg.1 |
TC1400006533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400144610.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144610.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144610.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144610.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144610.hg.1 |
TC1400006533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400144612.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144612.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144612.hg.1 |
ENST00000326783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400144612.hg.1 |
NM_022360.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400144612.hg.1 |
TC1400006533.hg.1 |
Clariom D TC |