Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019170.hg.1 |
ENST00000301061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019170.hg.1 |
ENST00000403957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019170.hg.1 |
ENST00000407467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019170.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019170.hg.1 |
TC12001464.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200180423.hg.1 |
TC1200010606.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000301061.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000403957.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000407467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000413630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000420388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
ENST00000475740.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010606.hg.1 |
NM_003394.3 |
Refseq |