Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011420.hg.1 |
TC05000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011420.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011420.hg.1 |
ENST00000359874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011420.hg.1 |
ENST00000398454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011420.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011420.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011420.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011419.hg.1 |
TC05000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011419.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011419.hg.1 |
ENST00000359874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011419.hg.1 |
ENST00000398454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011419.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011419.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011419.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000359874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000398454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000476697.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000507988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000508733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000521206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000359874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000398454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000476697.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000507988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000508733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
ENST00000521206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162841.hg.1 |
TC0500009031.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000359874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000398454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000476697.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000507988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000508733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000521206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000359874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000398454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000476697.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000507988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000508733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
ENST00000521206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162843.hg.1 |
TC0500009031.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000359874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000398454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000476697.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000507988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000508733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000521206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000256084.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000359874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000398454.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000476697.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000507988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000508733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
ENST00000521206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
NM_001127698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
NM_001127699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
NM_006846.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162842.hg.1 |
TC0500009031.hg.1 |
Clariom D TC |