Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014139.hg.1 |
ENST00000396060.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014139.hg.1 |
ENST00000396061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014139.hg.1 |
ENST00000396063.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014139.hg.1 |
ENST00000396064.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014139.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014139.hg.1 |
TC15001501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000472508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000472508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000472508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000267869.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000472508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000472508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000472508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000480768.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000484743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009621.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396060.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396063.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000396064.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000559141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
ENST00000559706.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
NM_004492.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500170900.hg.1 |
TC1500009621.hg.1 |
Clariom D TC |