Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009268.hg.1 |
TC04000642.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009268.hg.1 |
ENST00000281154.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009268.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159595.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159595.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159595.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159595.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159595.hg.1 |
TC0400008662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
ENST00000281154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008662.hg.1 |
NM_031291.3 |
Refseq |