Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222913.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222913.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222913.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200222913.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200222913.hg.1 |
TC0200016503.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
ENST00000361360.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016503.hg.1 |
NM_006236.2 |
Refseq |