Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256054.2 | NP_001242983.1 | 2 - 3 | 639 | 123 | |
NM_018325.4 | NP_060795.1 | 2 - 3 | 544 | 123 | ||
NM_145005.6 | NP_659442.2 | 2 - 3 | 561 | 123 | ||
GenBank mRNA | AK057806.1 | - | 2 - 3 | 539 | 123 | |
AK291425.1 | - | 2 - 3 | 534 | 123 | ||
AL832229.1 | - | 1 - 2 | 501 | 123 | ||
AL832467.1 | - | 2 - 3 | 500 | 123 | ||
BC020851.2 | - | 1 - 2 | 481 | 123 | ||
BC068445.1 | - | 1 - 2 | 493 | 5298741 | 123 | |
BG718065.1 | - | 2 - 3 | 550 | 123 | ||
BG722125.1 | - | 2 - 3 | 538 | 124 | ||
BQ068108.1 | - | 650 | 124 | |||
JN681271.1 | - | 2 - 3 | 617 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |