Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000556444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000556444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000557611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000557611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000557611.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012022.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012022.hg.1 |
ENST00000555449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012022.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012022.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012022.hg.1 |
TC14001051.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400166088.hg.1 |
TC1400008973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000331299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000546428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000555449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000556444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000556444.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000557611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000557611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000557611.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
ENST00000622765.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_001171170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008973.hg.1 |
NM_030631.3 |
Refseq |